Tisk…

Hydrogencitran diamonný, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Citric acid diammonium salt, Diammonium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate, Ammonium citrate dibasic

MQ200

247561-100G 247561-2.5KG 247561-500G
SIAL247561-100GEA 1110 CZK
SIAL247561-100G SIAL247561-2.5KG SIAL247561-500G
Hydrogencitran diamonný, Sigma-Aldrich®
Hydrogencitran diamonný
Vzorec: HOC(CO₂H)(CH₂CO₂NH₄)₂
MW: 226.19
Hustota: 1.483 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013068
CAS číslo: 3012-65-5
EINECS: 221-146-3
Merck Index: 13,00513

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR