Tisk…

Ethyl propionylacetate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

247162-5G
SIAL247162-5GEA 2710 CZK
SIAL247162-5G
Ethyl propionylacetate, Sigma-Aldrich®
Ethyl propionylacetate
Vzorec: C₇H₁₂O₃
MDL Number: MFCD00009317
CAS číslo: 4949-44-4
EINECS: 225-593-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR