Tisk…

N,N'-Bis(salicylidene)ethylenediamine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Ethylenebis(salicylimine), 2,2'-((Ethane-1,2-diylbis(azanylylidene))bis(methanylylidene))diphenol, SALEN

M-Clarity™ quality level = MQ100

236071-10G
SIAL236071-10GEA 1540 CZK
SIAL236071-10G
N,N'-Bis(salicylidene)ethylenediamine, Sigma-Aldrich®
N,N'-Bis(salicylidene)ethylenediamine
Vzorec: C₁₆H₁₆N₂O₂
MW: 268.31 g/mol
MDL Number: MFCD00002244
CAS číslo: 94-93-9
EINECS: 202-376-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR