Tisk…

N,N-Dimethyl-1,1,1-trimethylsilylamine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

226289-10G 226289-50G
SIAL226289-10GEA 1290 CZK
SIAL226289-10G SIAL226289-50G
N,N-Dimethyl-1,1,1-trimethylsilylamine, Sigma-Aldrich®
N,N-Dimethyl-1,1,1-trimethylsilylamine
Vzorec: C₅H₁₅NSi
Bod varu: 84 °C (1013 hPa)
Hustota: 0,7466 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008297
CAS číslo: 2083-91-2
EINECS: 218-222-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR