Tisk…

(-)-Quinine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (R)-[(2S,4S,5R)-1-Aza-5-vinylbicyclo[2.2.2]oct-2-yl](6-methoxyquinolin-4-yl)methanol, Quinine, (R)-(6-methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol

22620-100G 22620-25G 22620-5G
SIAL22620-100GEA 16700 CZK
SIAL22620-100G SIAL22620-25G SIAL22620-5G
(-)-Quinine, Sigma-Aldrich®
(-)-Quinine
Vzorec: C₂₀H₂₄N₂O₂
Bod tání: 176 °C
MDL Number: MFCD00198096
CAS číslo: 130-95-0
EINECS: 205-003-2
Merck Index: 12,08245

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR