Tisk…

Ammonium tetrafluoroborate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

223727-100G 223727-500G
SIAL223727-100GEA 908 CZK
SIAL223727-100G SIAL223727-500G
Ammonium tetrafluoroborate, Sigma-Aldrich®
Ammonium tetrafluoroborate
Vzorec: H₄BF₄N
Bod tání: 510 °C
Hustota: 1,87 g/cm³ (20 °C)
MDL Number: MFCD06245785
CAS číslo: 13826-83-0
EINECS: 237-531-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR