Tisk…

N-(1-Naphthyl)ethylenediamine dihydrochlorid, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: N-2-Aminoethyl-1-naftylamin dihydrochlorid

M-Clarity™ quality level = MQ200

222488-100G 222488-25G 222488-5G
SIAL222488-100GEA 24480 CZK
SIAL222488-100G SIAL222488-25G SIAL222488-5G
N-(1-Naphthyl)ethylenediamine dihydrochlorid, Sigma-Aldrich®
N-(1-Naphthyl)ethylenediamine dihydrochlorid
Vzorec: C₁₂H₁₆Cl₂N₂
Bod tání: 188…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00012556
CAS číslo: 1465-25-4
EINECS: 215-981-2
Merck Index: 12,06496
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3643e6df3e9cf8dc627823e9090c97004ee25308&vid=a30fc28279c11af12c6b735b00b84fc8f08caf34&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR