Tisk…

Trimethylsilylacetylene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

218170-1G 218170-25G 218170-5G 218170-100G
SIAL218170-1GEA 979 CZK
SIAL218170-1G SIAL218170-25G SIAL218170-5G SIAL218170-100G
Trimethylsilylacetylene, Sigma-Aldrich®
Trimethylsilylacetylene
Vzorec: C₅H₁₀Si
Bod varu: 53 °C (1013 hPa)
Hustota: 0,7111 g/cm³ (20 °C)
Bod vzplanutí: –34 °C
MDL Number: MFCD00008569
CAS číslo: 1066-54-2
EINECS: 213-919-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR