Tisk…

D-(-)-α-(4-Hydroxyphenyl)glycine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

215333-100G
SIAL215333-100GEA 0 CZK
SIAL215333-100G
D-(-)-α-(4-Hydroxyphenyl)glycine, Sigma-Aldrich®
D-(-)-α-(4-Hydroxyphenyl)glycine
Vzorec: C₈H₉NO₃
MDL Number: MFCD00004262
CAS číslo: 22818-40-2
EINECS: 245-247-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR