Tisk…

Cacodylic acid sodium salt trihydrate, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

20840-100G-F 20840-25G-F 20840-500G-F
SIAL20840-100G-FEA 6280 CZK
SIAL20840-100G-F SIAL20840-25G-F SIAL20840-500G-F
Cacodylic acid sodium salt trihydrate, Sigma-Aldrich®
Cacodylic acid sodium salt trihydrate
Vzorec: (CH₃)₂As(O)ONa·3H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149079
CAS číslo: 6131-99-3
EINECS: 204-708-2
UN: 1688
ADR: 6.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR