Tisk…

Diammonium tetrachloropalladate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

205850-2G
SIAL205850-2GEA 5660 CZK
SIAL205850-2G
Diammonium tetrachloropalladate, Sigma-Aldrich®
Diammonium tetrachloropalladate
Vzorec: H₈Cl₄N₂Pd
Hustota: 2,17 g/cm³ (20 °C)
MDL Number: MFCD00010884
CAS číslo: 13820-40-1
EINECS: 237-498-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR