Tisk…

Rhodium, powder, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

204218-1G
SIAL204218-1GEA 49070 CZK
SIAL204218-1G
Rhodium, powder, Sigma-Aldrich®
Rhodium
Vzorec: Rh
MDL Number: MFCD00011201
CAS číslo: 7440-16-6
EINECS: 231-125-0
UN: 3089
ADR: 4.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR