Tisk…

N-Benzylbenzamide, Sigma-Aldrich®

Dodavatel: Merck


MQ100

201014-5G
SIAL201014-5GEA 1040 CZK
SIAL201014-5G
N-Benzylbenzamide, Sigma-Aldrich®
N-Benzylbenzamide
Vzorec: C₁₄H₁₃NO
MW: 211.2591 g/mol
MDL Number: MFCD00003070
CAS číslo: 1485-70-7
EINECS: 216-063-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR