Tisk…

Trimethylaluminium 2 mol/l in toluene, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Trimethylaluminum

M-Clarity™ quality level = MQ200

198048-100ML 198048-1L 198048-800ML 198048-8L
SIAL198048-100MLEA 2850 CZK
SIAL198048-100ML SIAL198048-1L SIAL198048-800ML SIAL198048-8L
Trimethylaluminium 2 mol/l in toluene, Sigma-Aldrich®
Trimethylaluminium
Vzorec: C₃H₉Al
MDL Number: MFCD00008252
CAS číslo: 75-24-1
EINECS: 200-853-0
UN: 3399
ADR: 4.3,I

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR