Tisk…

Pentafluorobenzonitrile, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2,3,4,5,6-Pentafluorobenzonitrile

MQ100

196711-10G
SIAL196711-10GEA 1480 CZK
SIAL196711-10G
Pentafluorobenzonitrile, Sigma-Aldrich®
Pentafluorobenzonitrile
Vzorec: C₇F₅N
MW: 193.08
Bod varu: 185
Hustota: 1.566 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001775
CAS číslo: 773-82-0
EINECS: 212-259-9
UN: 1992
ADR: 3 III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR