Tisk…

D(+)-Biotin, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

19606-1G-F
SIAL19606-1G-FEA 9930 CZK
SIAL19606-1G-F
D(+)-Biotin, Sigma-Aldrich®
D(+)-Biotin
Vzorec: C₁₀H₁₆N₂O₃S
Bod tání: 232…233 °C
Hustota: 1,41 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00005541
CAS číslo: 58-85-5
EINECS: 200-399-3
Merck Index: 13,01231

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR