Tisk…

tert-Butylhydrazine hydrochloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

194972-100G 194972-500G 194972-5G
SIAL194972-100GEA 2370 CZK
SIAL194972-100G SIAL194972-500G SIAL194972-5G
tert-Butylhydrazine hydrochloride, Sigma-Aldrich®
tert-Butylhydrazine hydrochloride
Vzorec: C₄H₁₃ClN₂
MDL Number: MFCD00012947
CAS číslo: 7400-27-3
EINECS: 231-001-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR