Tisk…

4-Bromobenzyl alcohol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

187054-10G 187054-50G
SIAL187054-10GEA 1840 CZK
SIAL187054-10G SIAL187054-50G
4-Bromobenzyl alcohol, Sigma-Aldrich®
4-Bromobenzyl alcohol
Vzorec: C₇H₇BrO
Bod varu: 268 °C (1013 hPa)
Bod tání: 77…78 °C
MDL Number: MFCD00004650
CAS číslo: 873-75-6
EINECS: 212-851-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR