Tisk…

Dimethylsulfoxid-D6, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Dimethyl sulphoxide-[²H₆], Hexadeuterodimethyl sulphoxide, DMSO-D6, DMSO-d6, Dimethyl sulphoxide-d6

M-Clarity™ quality level = MQ100

185965-10G 185965-25G 185965-50G
SIAL185965-10GEA 2620 CZK
SIAL185965-10G SIAL185965-25G SIAL185965-50G
Dimethylsulfoxid-D6, Sigma-Aldrich®
Dimethylsulfoxid-D6
Vzorec: D₃CS(O)CD₃
Bod varu: 189 °C (5 mmHg)
Hustota: 1,19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002090
CAS číslo: 2206-27-1
EINECS: 218-617-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR