Tisk…

Ethyl-4-chloroacetoacetate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

180769-250G 180769-50G
SIAL180769-250GEA 3210 CZK
SIAL180769-250G SIAL180769-50G
Ethyl-4-chloroacetoacetate, Sigma-Aldrich®
Ethyl-4-chloroacetoacetate
Vzorec: C₆H₉ClO₃
MDL Number: MFCD00000939
CAS číslo: 638-07-3
EINECS: 211-317-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR