Tisk…

tert-Nonyl mercaptan (mixture of isomers) ≥97%, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: tert-Nonanethiol

M-Clarity™ quality level = MQ100

171034-250ML
SIAL171034-250MLEA 3340 CZK
SIAL171034-250ML
tert-Nonyl mercaptan (mixture of isomers) ≥97%, Sigma-Aldrich®
tert-Nonyl mercaptan(mixture)
Vzorec: C₉H₂₀S
MW: 160.32 g/mol
Bod varu: 188 °C (lit.)
Hustota: 0.856 g/ml
MDL Number: MFCD00059963
CAS číslo: 25360-10-5
UN: 3336
ADR: 3 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR