Tisk…

Dibenzyl azodicarboxylate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

166804-25G 166804-5G
SIAL166804-25GEA 9670 CZK
SIAL166804-25G SIAL166804-5G
Dibenzyl azodicarboxylate, Sigma-Aldrich®
Dibenzyl azodicarboxylate
Vzorec: C₁₆H₁₄N₂O₄
MDL Number: MFCD00016737
CAS číslo: 2449-05-0
EINECS: 219-508-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR