Tisk…

Trifluoromethanesulphonic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

158534-10G 158534-50G 158534-100G
SIAL158534-10GEA 1910 CZK
SIAL158534-10G SIAL158534-50G SIAL158534-100G
Trifluoromethanesulphonic acid, Sigma-Aldrich®
Trifluoromethanesulphonic acid
Vzorec: CHF₃O₃S
Bod varu: 162 °C (1013 hPa)
Bod tání: 35 °C
Hustota: 1,69 g/cm³ (20 °C)
MDL Number: MFCD00007514
CAS číslo: 1493-13-6
EINECS: 216-087-5
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR