Tisk…

2-Acetylbenzofuran, Sigma-Aldrich®

Dodavatel: Merck


MQ200

154377-25G
SIAL154377-25GEA 5690 CZK
SIAL154377-25G
2-Acetylbenzofuran, Sigma-Aldrich®
2-Acetylbenzofuran
Vzorec: C₁₀H₈O₂
MW: 160.17
MDL Number: MFCD00005849
CAS číslo: 1646-26-0
EINECS: 216-706-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR