Tisk…

Bismut(III) gallate basic hydrate ≥98% (by complexometric analysis, on dry substance), Sigma-Aldrich®

Dodavatel: Merck

15102-100G
SIAL15102-100GEA 1450 CZK
SIAL15102-100G
Bismut(III) gallate basic hydrate ≥98% (by complexometric analysis, on dry substance), Sigma-Aldrich®
Bismut(III) gallate basic hydrate
Vzorec: C₇H₅BiO₆·xH₂O
MDL Number: MFCD20441998
CAS číslo: 99-26-3
EINECS: 202-742-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR