Tisk…

(±)-Camphor-10-sulphonic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: DL-Camphor-10-sulphonic acid, (±)-2-Oxobornane-10-sulphonic acid, (7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonic acid

M-Clarity™ quality level = MQ200

147923-100G 147923-500G
SIAL147923-100GEA 1410 CZK
SIAL147923-100G SIAL147923-500G
(±)-Camphor-10-sulphonic acid, Sigma-Aldrich®
(±)-Camphor-10-sulphonic acid
Vzorec: C₁₀H₁₆O₄S
MW: 232.3
MDL Number: MFCD00074827
CAS číslo: 5872-08-2
EINECS: 227-527-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR