Tisk…

2-Chloro-5-nitrobenzaldehyde, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

139033-25G
SIAL139033-25GEA 3630 CZK
SIAL139033-25G
2-Chloro-5-nitrobenzaldehyde, Sigma-Aldrich®
2-Chloro-5-nitrobenzaldehyde
Vzorec: C₇H₄ClNO₃
MW: 185.57
Bod tání: 75…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00007293
CAS číslo: 6361-21-3
EINECS: 228-830-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR