Tisk…

1,8-Diazabicyclo[5.4.0]undec-7-ene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

139009-100G 139009-25G 139009-500G
SIAL139009-100GEA 5190 CZK
SIAL139009-100G SIAL139009-25G SIAL139009-500G
1,8-Diazabicyclo[5.4.0]undec-7-ene, Sigma-Aldrich®
1,8-Diazabicyclo[5.4.0]undec-7-ene
Vzorec: C₉H₁₆N₂
MDL Number: MFCD00006930
CAS číslo: 6674-22-2
EINECS: 229-713-7
UN: 3267
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR