Tisk…

trans-Cinnamic acid, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Cinnamic acid (only trans), (2E)-2-Phenyl-2-propenoic acid, trans-Cinnamic acid

M-Clarity™ quality level = MQ200

133760-100G 133760-2KG 133760-500G
SIAL133760-100GEA 899 CZK
SIAL133760-100G SIAL133760-2KG SIAL133760-500G
trans-Cinnamic acid, Sigma-Aldrich®
trans-Cinnamic acid
Vzorec: C₉H₈O₂
MW: 148.16
Bod varu: 300
Bod tání: 131…134 °C
Hustota: 1.25
MDL Number: MFCD00004369
CAS číslo: 140-10-3
EINECS: 205-398-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR