Tisk…

2-Furohydrazide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

130443-5G
SIAL130443-5GEA 1050 CZK
SIAL130443-5G
2-Furohydrazide, Sigma-Aldrich®
2-Furohydrazide
Vzorec: C₅H₆N₂O₂
MDL Number: MFCD00003235
CAS číslo: 3326-71-4
EINECS: 222-046-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR