Tisk…

N,O-Bis(trimethylsilyl)acetamid, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: BSA, Silyl Derivatisation, Trimethylsilyl N-(trimethylsilyl)acetimidate, Trimethylsilyl N-trimethylsilylacetamidate

M-Clarity™ quality level = MQ100

128910-100ML 128910-10ML 128910-1L 128910-25ML
SIAL128910-100MLEA 4870 CZK
SIAL128910-100ML SIAL128910-10ML SIAL128910-1L SIAL128910-25ML
N,O-Bis(trimethylsilyl)acetamid, Sigma-Aldrich®
N,O-Bis(trimethylsilyl)acetamid
Vzorec: C₈H₂₁NOSi₂
MW: 203.43
Bod varu: 162
Bod tání: ∼24 °C
Hustota: 0.82 g/cm³ (20 °C)
Bod vzplanutí: 40
MDL Number: MFCD00008270
CAS číslo: 10416-59-8
EINECS: 233-892-7
UN: 2920
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR