Tisk…

3-Chlorobenzylamine, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: (3-Chlorophenyl)methanamine

M-Clarity™ quality level = MQ100

127167-5G
SIAL127167-5GEA 2460 CZK
SIAL127167-5G
3-Chlorobenzylamine, Sigma-Aldrich®
3-Chlorobenzylamine
Vzorec: C₇H₈ClN
MW: 141,6 g/mol
Bod varu: 40 °C (0,8 mmHg)
Hustota: 1,159 g/cm³ (20 °C)
MDL Number: MFCD00040752
CAS číslo: 4152-90-3
EINECS: 223-985-0
UN: 2735
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR