Tisk…

Diethyl azelate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

124583-100ML
SIAL124583-100MLEA 14360 CZK
SIAL124583-100ML
Diethyl azelate, Sigma-Aldrich®
Diethyl azelate
Vzorec: C₁₃H₂₄O₄
MDL Number: MFCD00009218
CAS číslo: 624-17-9
EINECS: 210-833-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR