Tisk…

2-Amino-4-methylpyridine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Aminopicoline, 2-Amino-4-picoline, 4-Methyl-2-pyridylamine, 4-Methylpyridin-2-amine

M-Clarity™ quality level = MQ100

123080-100G
SIAL123080-100GEA 1690 CZK
SIAL123080-100G
2-Amino-4-methylpyridine, Sigma-Aldrich®
2-Amino-4-methylpyridine
Vzorec: C₆H₈N₂
MW: 108.14
Bod varu: 230.9
Bod tání: 98
Storage Temperature: Ambient
MDL Number: MFCD00006321
CAS číslo: 695-34-1
EINECS: 211-780-9
UN: 2811
ADR: 6.1 III
Merck Index: 13,00465

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR