Tisk…

Bis(8-hydroxyquinolinium)sulphate monohydrate, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Quinolin-8-ol sulphate hydrate, Quinolin-8-ol hemisulphate hemihydrate, 8-Hydroxyquinoline sulphate monohydrate, 8-Hydroxyquinolinium hemisulphate hemihydrate, 8-Hydroxyquinoline hemisulphate salt hemihydrate

M-Clarity™ quality level = MQ200

108073-100G
SIAL108073-100GEA 2060 CZK
SIAL108073-100G
Bis(8-hydroxyquinolinium)sulphate monohydrate, Sigma-Aldrich®
Bis(8-hydroxyquinolinium)sulphate monohydrate
Vzorec: C₁₈H₁₆N₂O₆S·1H₂O
MDL Number: MFCD00149610
CAS číslo: 207386-91-2
EINECS: 205-137-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR