Tisk…

2-Iodohippuric acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: N-(2-Iodobenzoyl)glycine

M-Clarity™ quality level = MQ100

106577-10G
SIAL106577-10GEA 2890 CZK
SIAL106577-10G
2-Iodohippuric acid, Sigma-Aldrich®
2-Iodohippuric acid
Vzorec: C₉H₈INO₃
MW: 305,07 g/mol
MDL Number: MFCD00002694
CAS číslo: 147-58-0
EINECS: 205-693-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR