Tisk…

Dimethoxy(dimethyl)silane, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

104906-1L 104906-500ML
SIAL104906-1LEA 4790 CZK
SIAL104906-1L SIAL104906-500ML
Dimethoxy(dimethyl)silane, Sigma-Aldrich®
Dimethoxy(dimethyl)silane
Vzorec: C₄H₁₂O₂Si
Bod varu: 82 °C (1013 hPa)
Bod tání: –80 °C
Hustota: 0,8638 g/cm³ (20 °C)
MDL Number: MFCD00025691
CAS číslo: 1112-39-6
EINECS: 214-189-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR