Tisk…

Pentafluorobenzyl bromide, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: α-Bromo-2,3,4,5,6-pentafluorotoluene, 2,3,4,5,6-Pentafluorobenzyl bromide

M-Clarity™ quality level = MQ200

101052-5G 101052-1G 101052-25G
SIAL101052-5GEA 2330 CZK
SIAL101052-5G SIAL101052-1G SIAL101052-25G
Pentafluorobenzyl bromide, Sigma-Aldrich®
Pentafluorobenzyl bromide
Vzorec: (C₆F₅)CH₂Br
MW: 260.99
Bod varu: 174
Hustota: 1.72
Storage Temperature: Ambient
MDL Number: MFCD00000299
CAS číslo: 1765-40-8
EINECS: 217-182-4
UN: 3265
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR