Tisk…

trans-4-Fluoro-β-nitrostyrene, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Fluoro-β-nitrostyrene

M-Clarity™ quality level = MQ100

09506-5G
SIAL09506-5GEA 2950 CZK
SIAL09506-5G
trans-4-Fluoro-β-nitrostyrene, Sigma-Aldrich®
trans-4-Fluoro-β-nitrostyrene
Vzorec: C₈H₆FNO₂
MW: 167,14 g/mol
MDL Number: MFCD00024824
CAS číslo: 706-08-1
EINECS: 211-891-2
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR