Tisk…

1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1-Butyl-2,3-dimethylimidazolium Tetrafluoroborate, [BDiMIM] [BF4]

M-Clarity™ quality level = MQ100

04383-50G-F
SIAL04383-50G-FEA 12550 CZK
SIAL04383-50G-F
1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate, Sigma-Aldrich®
1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate
Vzorec: C₉H₁₇BF₄N₂
MW: 240.05
Hustota: 1.2
MDL Number: MFCD03427618
CAS číslo: 402846-78-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR