Tisk…

Acetylthiocholinjodid, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ100

01480-1G 01480-25G 01480-5G
SIAL01480-1GEA 1650 CZK
SIAL01480-1G SIAL01480-25G SIAL01480-5G
Acetylthiocholinjodid, Sigma-Aldrich®
Acetylthiocholinjodid
Vzorec: H₃CC(S)O(CH₂)₂N(CH₃)₃I
Bod tání: 204 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011819
CAS číslo: 1866-15-5
EINECS: 217-474-1
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9024633662ddbbc8a8efbcd594cd79dbbcbb2d6&vid=c8a063af769022ab97d866f5f7547b476c5949a7&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR