Tisk…
Shodex HILICpak VN-50 HILIC Columns for Oligonucleotides and Oligosaccharides
Dodavatel: Resonac
F6711400 F6711500 F7630500 F7630600 F6830100
SHOWF6711400EA 17100 CZK
SHOWF6711400 SHOWF6711500 SHOWF7630500 SHOWF7630600 SHOWF6830100
Shodex HILICpak VN-50 HILIC Columns for Oligonucleotides and Oligosaccharides
Kolony chromatografické
Robust polymer-based HILIC column for separation by size/chain length.
  • Polymer-based HILIC columns with diol (-OH) groups
  • Best for oligosaccharides and oligonucleotides
  • Suitable for oligomer separation which is not possible by SEC column or conventional HILIC columns
  • The modified diol groups on the packing material create the HILIC mode
  • Polymer-based packing material provides excellent chemical stability and minimum deterioration over an extended time period
  • pH stability: 2 to 13
  • Also suitable for evaporative light scattering detector (ELSD), corona
More Product Information