Tisk…
Horse Recombinant RE VEGF-165 (Animal free) (from E. coli)
Dodavatel: Shenandoah Biotechnology
400-18AF-100UG 400-18AF-1MG 400-18AF-20UG 400-18AF-500UG 400-18AF-5UG
SHBT400-18AF-100UGEA 15990 CZK
SHBT400-18AF-100UG SHBT400-18AF-1MG SHBT400-18AF-20UG SHBT400-18AF-500UG SHBT400-18AF-5UG
Horse Recombinant RE VEGF-165 (Animal free) (from E. coli)
Proteiny
FLT-3 Ligand is a growth factor that regulates proliferation of early hematopoietic cells. FLT-3 Ligand binds to cells expressing the tyrosine kinase receptor FLT-3. FLT-3 ligand by itself does not stimulate proliferation of early hematopoietic cells, but synergizes with other CSFs and interleukins to induce growth and differentiation. Unlike SCF, FLT-3 ligand exerts no activity on mast cells. Multiple isoforms of FLT-3 Ligand have been identified.
  • High quality
  • Low endotoxin
  • Affordable
The predominant biologically active form is anchored to the cell surface as the extracellular domain of a transmembrane protein (209 aa). The membrane-bound isoform can be proteolytically cleaved to generate a biologically active soluble isoform. The biological activity of CTGrade GMP interleukins and growth factors is standardized, where applicable, to WHO International standards, providing cell and gene therapy developers consistent, lot-to-lot biological activity and performance.
Upozornení: For research use or further manufacturing only.