Tisk…

Pig Recombinant IL-4 (Animal free) (from E. coli)

Dodavatel: Shenandoah Biotechnology

400-10AF-5UG 400-10AF-20UG 400-10AF-100UG 400-10AF-500UG 400-10AF-1MG
SHBT400-10AF-5UGEA 2230 CZK
SHBT400-10AF-5UG SHBT400-10AF-20UG SHBT400-10AF-100UG SHBT400-10AF-500UG SHBT400-10AF-1MG
Pig Recombinant IL-4 (Animal free) (from E. coli)
Proteiny

Granulocyte-macrophage colony-stimulating factor (GM-CSF) is a hematopoietic growth factor produced by endothelial cells, monocytes, fibroblasts, and T cells. GM-CSF stimulates the production of neutrophilic granulocytes, macrophages, and mixed granulocyte-macrophage colonies from bone marrow cells. GM-CSF promotes immune system development and regulates neutrophil function during infection.


  • High quality
  • Low endotoxin
  • Affordable


Informace o dodávce: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.


Upozornení: For research use only.

Objednat

Produktová specifikace

Název proteinu/peptidu IL-4
Environmentally Preferable
Proteinová synonyma BCGF|B-cell stimulatory factor-1|BCDF|BSF-1
Typ proteinu/peptidu Recombinant
Proteinový podtyp Cytokine
Druhy Pig
Zdroj E. coli
Spojení Unconjugated
Identifikační číslo genu Q04745
Sekvence MHKCDITLQE IIKTLNILTA RKNSCMELPV TDVFAAPENT TEKETFCRAS TVLRHIYRHH TCMKSLLSGL DRNLSSMANM TCSVHEAKKS TLKDFLERLK TIMKEKYSKC
Bez obsahu proteázy N
Bez obsahu živočichů Y
Bez nukleáz N
Bez nosiče Y
Bez obsahu endotoxinů N
Úroveň endotoxinů Low
Obsah endotoxinů Endotoxin LAL (EU/µg) ≤0.1
UniProtKB Q04745
Biologická aktivita Bioactivity assay: TF-1 cell proliferation, ED50 ≤2 ng/ml, bioactivity ≥5,0×10⁵ units/mg
Čistota ≥95%
Třída RUO
Formulace 10 mM sodium phosphate, pH 7,5
Molekulární hmotnost 12,7 kDa
Pokyny pro rekonstituce Sterile water at 0,1 mg/ml
Podmínky uskladnění −20 °C
Transportní teplota RT
Testované aplikace Bioassay

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR