Tisk…

Rat Recombinant SHH (from E. coli)

Dodavatel: Shenandoah Biotechnology

300-40-5UG 300-40-25UG 300-40-100UG 300-40-500UG 300-40-1MG
SHBT300-40-5UGEA 2070 CZK
SHBT300-40-5UG SHBT300-40-25UG SHBT300-40-100UG SHBT300-40-500UG SHBT300-40-1MG
Rat Recombinant SHH (from E. coli)
Proteiny

Sonic hedgehog (SHH) is a member of a small group of hedgehog secreted proteins that are essential for development in both vertebrates and invertebrates. There are three mammalian hedgehog homologues, sonic, desert, and indian, that signal via the Patched-1 and Patched-2 receptors. SHH is a morphogen that is essential during vertebrate organogenesis and adult stem cell division.


  • High quality
  • Low endotoxin
  • Affordable


There are three mammalian hedgehog homologues, sonic, desert, and indian, that signal via the Patched-1 and Patched-2 receptors. SHH is a morphogen that is essential during vertebrate organogenesis and adult stem cell division.


Informace o dodávce: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.


Upozornení: For research use only.

Objednat

Produktová specifikace

Název proteinu/peptidu SHH
Environmentally Preferable
Proteinová synonyma Shh|HHG-1
Typ proteinu/peptidu Recombinant
Proteinový podtyp Cytokine
Druhy Rat
Zdroj E. coli
Spojení Unconjugated
Identifikační číslo genu Q63673
Sekvence MIIGPGRGFG KRQHPKKLTP LAYKQFIPNV AEKTLGASGR YEGKITRNSE RFKELTPNYN PDIIFKDEEN TGADRLMTQR CKDKLNALAI SVMNQWPGVK LRVTEGWDED GHHSEESLHY EGRAVDITTS DRDRSKYGML ARLAVEAGFD WVYYESKARI HCSVKAENSV AAKSDG
Bez obsahu proteázy N
Bez obsahu živočichů N
Bez nukleáz N
Bez nosiče Y
Bez obsahu endotoxinů N
Úroveň endotoxinů Low
Obsah endotoxinů Endotoxin LAL (EU/µg) ≤0.1
UniProtKB Q63673
Biologická aktivita Bioactivity assay: Currently undetermined, ED50 ≤Currently undetermined, bioactivity ≥NA
Čistota ≥95%
Třída RUO
Formulace 10 mM sodium phosphate, pH 7,5
Molekulární hmotnost 19,9 kDa
Pokyny pro rekonstituce Sterile water at 0,1 mg/ml
Podmínky uskladnění −20 °C
Transportní teplota RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR