Tisk…
Mouse Recombinant MCP-1 / CCL2 (from E. coli)
Dodavatel: Shenandoah Biotechnology
200-39-100UG 200-39-10UG 200-39-1MG 200-39-500UG
SHBT200-39-100UGEA 12920 CZK
SHBT200-39-100UG SHBT200-39-10UG SHBT200-39-1MG SHBT200-39-500UG
Mouse Recombinant MCP-1 / CCL2 (from E. coli)
Proteiny
Monocyte chemotactic protein 1 (MCP-1), also known as CCL2, is produced by injured or infected tissues. MCP-1 signals through the CCR2 and CCR4 G protein-coupled receptors to recruit memory T cells, monocytes, and dendritic cells to sites of inflammation.
  • High Quality
  • Low Endotoxin
  • Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Upozornení: For research use only.