Tisk…
Mouse Recombinant IFN-G (Animal free) (from E. coli)
Dodavatel: Shenandoah Biotechnology
200-16AF-100UG 200-16AF-1MG 200-16AF-20UG 200-16AF-250UG 200-16AF-500UG
SHBT200-16AF-100UGEA 5680 CZK
SHBT200-16AF-100UG SHBT200-16AF-1MG SHBT200-16AF-20UG SHBT200-16AF-250UG SHBT200-16AF-500UG
Mouse Recombinant IFN-G (Animal free) (from E. coli)
Proteiny
Interleukin 5 (IL-5) is a hematopoietic growth factor that is expressed in type 2 T helper (Th2) cells, mast cells, and eosinophils. IL-5 acts through the IL-5 receptor (IL-5R), stimulates B cell growth, and mediates eosinophil activation. IL-5 expression is regulated by the GATA binding protein 3 (GATA3) transcription factor. Human and mouse IL-5 show cross-reactivity.
  • High quality
  • Low endotoxin
  • Affordable
Interferon gamma (IFN-γ) is a type II interferon that is critical during adaptive and innate immune responses to infection. IFN-γ is produced by T cells and natural killer cells following antigen-specific activation. IFN-γ binds IFN-γ receptors (IFN-γ R1 and IFN-γ R2), which are expressed on most immune cells, to activate the JAK-STAT pathway. IFN-γ-induced signaling increases the expression of class 1 major histocompatibility complex (MHC) molecules. Mouse IFN-γ is not cross-reactive with human IFN-γ. 
Informace o dodávce: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Upozornení: For research use only.