Tisk…
Mouse Recombinant IFN-G (from E. coli)
Dodavatel: Shenandoah Biotechnology
200-16-100UG 200-16-1MG 200-16-20UG 200-16-250UG 200-16-500UG
SHBT200-16-100UGEA 5160 CZK
SHBT200-16-100UG SHBT200-16-1MG SHBT200-16-20UG SHBT200-16-250UG SHBT200-16-500UG
Mouse Recombinant IFN-G (from E. coli)
Proteiny
Interferon gamma (IFN-γ) is a type II interferon that is critical during adaptive and innate immune responses to infection. IFN-γ is produced by T cells and natural killer cells following antigen-specific activation. IFN-γ binds IFN-γ receptors (IFN-γ R1 and IFN-γ R2), which are expressed on most immune cells, to activate the JAK-STAT pathway. IFN-γ-induced signaling increases the expression of class 1 major histocompatibility complex (MHC) molecules. Mouse IFN-γ is not cross-reactive with human IFN-γ.
  • High quality
  • Low endotoxin
  • Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Informace o dodávce: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Upozornení: For research use only.