Tisk…
Human Recombinant TNFR-1 (from E. coli)
Dodavatel: Shenandoah Biotechnology
100-35-100UG 100-35-1MG 100-35-20UG 100-35-500UG 100-35-5UG
SHBT100-35-100UGEA 12920 CZK
SHBT100-35-100UG SHBT100-35-1MG SHBT100-35-20UG SHBT100-35-500UG SHBT100-35-5UG
Human Recombinant TNFR-1 (from E. coli)
Proteiny
Tumor necrosis factor receptor type 1 (TNFR-1) is expressed in most tissues and is activated by soluble and membrane-bound tumor necrosis factor alpha (TNF-α). TNFR-1 activates NF-κB and MAPK pathways to induce inflammation, promote apoptotic cell death, inhibit tumorigenesis, and inhibit viral replication.
  • High quality
  • Low endotoxin
  • Affordable
Upozornení: For research use only.