Tisk…
Human Recombinant MIG/CXCL9 (from E. coli)
Dodavatel: Shenandoah Biotechnology
100-184-100UG 100-184-1MG 100-184-20UG 100-184-500UG 100-184-5UG
SHBT100-184-100UGEA 12920 CZK
SHBT100-184-100UG SHBT100-184-1MG SHBT100-184-20UG SHBT100-184-500UG SHBT100-184-5UG
Human Recombinant MIG/CXCL9 (from E. coli)
Proteiny
Platelet-derived growth factor (PDGF) is an important regulator of cell growth, proliferation, and angiogenesis. PDGF synthesis is induced by IL-1, IL-6, TNF-α, TGF-β, and EGF signaling. PDGF functions as a mitogenic growth hormone on cells of mesenchymal lineage, such as smooth muscle and glial cells.
  • High quality
  • Low endotoxin
  • Affordable
PDGF is also stored in the alpha-granules of platelets and is released upon adherence to traumatized tissues. PDGF is a dimeric glycoprotein formed by two A chains (AA), two B chains (BB), or as a heterodimer with an A and a B chain (AB). The PDGF dimer binds the cell surface receptor tyrosine kinases PDGFR-α and PDGFR-β.
Informace o dodávce: A Certificate of Analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Upozornení: For research use only.